Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1-(4-((4-(4-Hydroxypiperidin-1-yl)phenyl)amino)-5-oxo-5,6-dihydropyrimido[4,5-d]pyridazin-2-yl)piperidin-4-yl)acetonitrile 99.41%

Supplier: AMBEED, INC
Danger

A1176575-50MG A1176575-25MG
76676-388EA 242 USD InStock
76676-388 77620-494
2-(1-(4-((4-(4-Hydroxypiperidin-1-yl)phenyl)amino)-5-oxo-5,6-dihydropyrimido[4,5-d]pyridazin-2-yl)piperidin-4-yl)acetonitrile 99.41%
2-(1-(4-((4-(4-Hydroxypiperidin-1-yl)phenyl)amino)-5-oxo-5,6-dihydropyrimido[4,5-d]pyridazin-2-yl)piperidin-4-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₂₄H₂₈N₈O₂
MW: 460.53 g/mol
MDL Number: MFCD31692335
CAS Number: 1425381-60-7
UN: 3077
ADR: 9 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty