Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

SBI-115 99%

Supplier: AMBEED, INC
Warning

A1176731-1MG A1176731-50MG A1176731-5MG
76676-400EA 43 USD InStock
76676-400 76676-402 76676-404
SBI-115 99%
SBI-115
Precaución: for Research Use Only


Formula: C₁₄H₁₃ClN₂O₄S
MDL Number: MFCD07537427
CAS Number: 882366-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty