Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-((5-(3-Benzyl-5-hydroxy-4-oxo-1,2,3,4-tetrahydroquinazolin-2-yl)-2-methoxybenzyl)oxy)phenyl)acetamide 98%

Supplier: AMBEED, INC
Warning

A1176977-10MG A1176977-1MG A1176977-5MG
76676-434EA 113 USD InStock
76676-434 76676-436 76676-438
N-(4-((5-(3-Benzyl-5-hydroxy-4-oxo-1,2,3,4-tetrahydroquinazolin-2-yl)-2-methoxybenzyl)oxy)phenyl)acetamide 98%
N-(4-((5-(3-Benzyl-5-hydroxy-4-oxo-1,2,3,4-tetrahydroquinazolin-2-yl)-2-methoxybenzyl)oxy)phenyl)acetamide
Precaución: for Research Use Only


Formula: C₃₁H₂₉N₃O₅
MDL Number: MFCD31746908
CAS Number: 1186649-91-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty