Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-Tetrahydrofuran-3-yl 4-(6-(5-(4-ethoxy-1-isopropylpiperidin-4-yl)pyridin-2-yl)pyrrolo[1,2-b]pyridazin-4-yl)piperazine-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A1177054-10MG A1177054-1MG A1177054-25MG A1177054-50MG A1177054-5MG
76676-454EA 163 USD InStock
76676-454 76676-456 76676-458 76676-460 76676-462
(R)-Tetrahydrofuran-3-yl 4-(6-(5-(4-ethoxy-1-isopropylpiperidin-4-yl)pyridin-2-yl)pyrrolo[1,2-b]pyridazin-4-yl)piperazine-1-carboxylate 98%
(R)-Tetrahydrofuran-3-yl 4-(6-(5-(4-ethoxy-1-isopropylpiperidin-4-yl)pyridin-2-yl)pyrrolo[1,2-b]pyridazin-4-yl)piperazine-1-carboxylate
Precaución: for Research Use Only


Formula: C₃₁H₄₂N₆O₄
MW: 562.7030 g/mol
MDL Number: MFCD32062780
CAS Number: 2141955-96-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty