Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-4-fluorophenyl)(4-fluoro-4-((((5-methylpyrimidin-2-yl)methyl)amino)methyl)piperidin-1-yl)methanone ≥98%

Supplier: AMBEED, INC
Warning

A1177359-10MG A1177359-1MG A1177359-25MG A1177359-5MG
76676-480EA 320 USD InStock
76676-480 76676-482 76676-484 76676-486
(3-Chloro-4-fluorophenyl)(4-fluoro-4-((((5-methylpyrimidin-2-yl)methyl)amino)methyl)piperidin-1-yl)methanone ≥98%
(3-Chloro-4-fluorophenyl)(4-fluoro-4-((((5-methylpyrimidin-2-yl)methyl)amino)methyl)piperidin-1-yl)methanone
Precaución: for Research Use Only


Formula: C₁₉H₂₁ClF₂N₄O
MDL Number: MFCD28502284
CAS Number: 635323-95-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty