Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 7-chloro-3,4-dihydro-1,8-naphthyridine-1(2H)-carboxylate 98%

Supplier: AMBEED, INC
Danger

A1181404-100MG A1181404-1G A1181404-250MG A1181404-5G A1181404-25G
76676-708EA 5 USD InStock
76676-708 76676-710 76676-712 76676-714 77620-640
tert-Butyl 7-chloro-3,4-dihydro-1,8-naphthyridine-1(2H)-carboxylate 98%
tert-Butyl 7-chloro-3,4-dihydro-1,8-naphthyridine-1(2H)-carboxylate
Precaución: for Research Use Only


Formula: C₁₃H₁₇ClN₂O₂
MDL Number: MFCD16036500
CAS Number: 679392-23-5
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty