Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (R)-2-bromobutanoate 98%

Supplier: AMBEED, INC
Danger

A1186683-1G A1186683-250MG A1186683-5G
76676-718EA 36 USD InStock
76676-718 76676-720 76676-722
Methyl (R)-2-bromobutanoate 98%
Methyl (R)-2-bromobutanoate
Precaución: for Research Use Only


Formula: C₅H₉BrO₂
MDL Number: MFCD20622163
CAS Number: 114438-75-4
UN: 2924
ADR: 3,8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty