Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4'-(Anthracen-9-yl)-2,2':6',2''-terpyridine 98%

Supplier: AMBEED, INC
Warning

A1186735-1G A1186735-100MG A1186735-250MG
76676-728EA 122 USD InStock
76676-728 77620-666 77620-668
4'-(Anthracen-9-yl)-2,2':6',2''-terpyridine 98%
4'-(Anthracen-9-yl)-2,2':6',2''-terpyridine
Precaución: for Research Use Only


Formula: C₂₉H₁₉N₃
MW: 409.4813 g/mol
MDL Number: MFCD14636247
CAS Number: 163087-28-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty