Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-(5-methoxy-1H-indol-3-yl)propanoate 97+%

Supplier: AMBEED, INC
Warning

A1187875-100MG A1187875-1G A1187875-250MG
76676-744EA 13 USD InStock
76676-744 76676-746 76676-748
Ethyl 3-(5-methoxy-1H-indol-3-yl)propanoate 97+%
Ethyl 3-(5-methoxy-1H-indol-3-yl)propanoate
Precaución: for Research Use Only


Formula: C₁₄H₁₇NO₃
MDL Number: MFCD31716142
CAS Number: 76834-78-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty