Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-(2-Methoxy-6H-dipyrido[2,3-a:3',2'-e]pyrrolizin-11-yl)ethyl)furan-2-carboxamide 98+%

Supplier: AMBEED, INC
Warning

A1191668-10MG A1191668-1MG A1191668-5MG
76676-960EA 62 USD InStock
76676-960 76676-962 76676-964
N-(2-(2-Methoxy-6H-dipyrido[2,3-a:3',2'-e]pyrrolizin-11-yl)ethyl)furan-2-carboxamide 98+%
N-(2-(2-Methoxy-6H-dipyrido[2,3-a:3',2'-e]pyrrolizin-11-yl)ethyl)furan-2-carboxamide
Precaución: for Research Use Only


Formula: C₂₁H₁₈N₄O₃
MDL Number: MFCD28160701
CAS Number: 874484-20-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty