Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(2,5-dioxopyrrolidin-1-yl) (disulfanediylbis(ethane-2,1-diyl)) dicarbonate 97%

Supplier: AMBEED, INC
Warning

A1192322-100MG A1192322-1G A1192322-250MG
76677-016EA 91 USD InStock
76677-016 76677-018 76677-020
Bis(2,5-dioxopyrrolidin-1-yl) (disulfanediylbis(ethane-2,1-diyl)) dicarbonate 97%
Bis(2,5-dioxopyrrolidin-1-yl) (disulfanediylbis(ethane-2,1-diyl)) dicarbonate
Precaución: for Research Use Only


Formula: C₁₄H₁₆N₂O₁₀S₂
MDL Number: MFCD32644554
CAS Number: 1688598-83-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty