Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalaldehyde 98%

Supplier: AMBEED, INC
Warning

A1193144-100MG A1193144-1G A1193144-250MG A1193144-5G
76677-056EA 10 USD InStock
76677-056 76677-058 76677-060 76677-062
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalaldehyde 98%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isophthalaldehyde
Precaución: for Research Use Only


Formula: C₁₄H₁₇BO₄
MW: 260.0934 g/mol
MDL Number: MFCD22493812
CAS Number: 945865-80-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty