Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dianiline 98%

Supplier: AMBEED, INC
Warning

A1193176-100MG A1193176-1G A1193176-250MG A1193176-5G
76677-104EA 18 USD InStock
76677-104 76677-106 76677-108 76677-110
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dianiline 98%
4,4'-(Benzo[c][1,2,5]thiadiazole-4,7-diyl)dianiline
Precaución: for Research Use Only


Formula: C₁₈H₁₄N₄S
MDL Number: MFCD33023608
CAS Number: 1203707-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty