Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-4-(trifluoromethyl)-2,3-dihydro-1H-inden-1-one 97%

Supplier: AMBEED, INC
Warning

A1195014-100MG A1195014-1G A1195014-250MG
76677-294EA 40 USD InStock
76677-294 76677-296 76677-298
6-Bromo-4-(trifluoromethyl)-2,3-dihydro-1H-inden-1-one 97%
6-Bromo-4-(trifluoromethyl)-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only


Formula: C₁₀H₆BrF₃O
MW: 279.0533 g/mol
MDL Number: MFCD18647940
CAS Number: 1273655-84-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty