Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Naphthalene-2,6-dicarbonyl dichloride 95%

Supplier: AMBEED, INC
Danger

A1195881-10G A1195881-1G A1195881-25G A1195881-5G
76677-416EA 143 USD InStock
76677-416 76677-418 76677-420 76677-422
Naphthalene-2,6-dicarbonyl dichloride 95%
Naphthalene-2,6-dicarbonyl dichloride
Precaución: for Research Use Only


Formula: C₁₂H₆Cl₂O₂
MW: 253.0808 g/mol
MDL Number: MFCD00228481
CAS Number: 2351-36-2
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty