Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2,3,6-Trifluorophenyl)hydrazine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A1197035-100MG A1197035-1G
76677-494EA 97 USD InStock
76677-494 76677-496
(2,3,6-Trifluorophenyl)hydrazine hydrochloride 98%
(2,3,6-Trifluorophenyl)hydrazine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₆ClF₃N₂
MDL Number: MFCD31421786
CAS Number: 2137836-61-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty