Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

22-Methoxy-22-oxodocosanoic acid 98%

Supplier: AMBEED, INC
Warning

A1204793-1G A1204793-250MG
76677-542EA 38 USD InStock
76677-542 76677-544
22-Methoxy-22-oxodocosanoic acid 98%
22-Methoxy-22-oxodocosanoic acid
Precaución: For research use only.


Formula: C₂₃H₄₄O₄
MW: 384.5931 g/mol
CAS Number: 7511-30-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty