Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,5-dimethylphenyl)-6-isopropylisoquinoline 98%

Supplier: AMBEED, INC
Warning

A1206079-1G A1206079-250MG A1206079-5G
76677-626EA 15 USD InStock
76677-626 76677-628 76677-630
1-(3,5-dimethylphenyl)-6-isopropylisoquinoline 98%
1-(3,5-dimethylphenyl)-6-isopropylisoquinoline
Precaución: for Research Use Only


Formula: C₂₀H₂₁N
MDL Number: MFCD32067793
CAS Number: 1936437-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty