Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitrobenzo[c][1,2]oxaborol-1(3H)-ol 98%

Supplier: AMBEED, INC
Warning

A1207481-1G A1207481-250MG A1207481-5G
76677-762EA 211 USD InStock
76677-762 76677-764 76677-766
5-Nitrobenzo[c][1,2]oxaborol-1(3H)-ol 98%
5-Nitrobenzo[c][1,2]oxaborol-1(3H)-ol
Precaución: for Research Use Only


Formula: C₇H₆BNO₄
MDL Number: MFCD24857368
CAS Number: 875816-94-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty