Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(tert-Butyl)-4-(2,6-diisopropoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%

Supplier: AMBEED, INC
Warning

A1207631-100MG A1207631-10MG A1207631-250MG
76677-776EA 152 USD InStock
76677-776 77620-828 77620-830
(S)-3-(tert-Butyl)-4-(2,6-diisopropoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 97%
(S)-3-(tert-Butyl)-4-(2,6-diisopropoxyphenyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole
Precaución: for Research Use Only


Formula: C₂₃H₃₁O₃P
MDL Number: MFCD32641148
CAS Number: 2448341-44-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty