Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

rel-(2R,3R)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-(2,4,6-triisopropylbenzyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 98%

Supplier: AMBEED, INC
Warning

A1207670-100MG A1207670-250MG A1207670-50MG
76677-844EA 157 USD InStock
76677-844 77620-836 77620-838
rel-(2R,3R)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-(2,4,6-triisopropylbenzyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole 98%
rel-(2R,3R)-3-(tert-Butyl)-4-(2,6-dimethoxyphenyl)-2-(2,4,6-triisopropylbenzyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole
Precaución: for Research Use Only


Formula: C₃₅H₄₇O₃P
CAS Number: 1908442-13-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty