Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-chloro-5-nitrobenzoic acid 97%

Supplier: AMBEED, INC
Warning

A1207983-100MG A1207983-250MG
76677-868EA 14 USD InStock
76677-868 76677-870
2-Bromo-3-chloro-5-nitrobenzoic acid 97%
2-Bromo-3-chloro-5-nitrobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₃BrClNO₄
MDL Number: MFCD24108588
CAS Number: 1499553-66-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty