Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2S)-5-(2-Chloroethanimidamido)-2-(phenylformamido)pentanamide trifluoroacetic acid salt 98%

Supplier: AMBEED, INC
Warning

A1209276-10MG A1209276-1MG A1209276-5MG
76677-924EA 77 USD InStock
76677-924 76677-926 76677-928
(2S)-5-(2-Chloroethanimidamido)-2-(phenylformamido)pentanamide trifluoroacetic acid salt 98%
(2S)-5-(2-Chloroethanimidamido)-2-(phenylformamido)pentanamide trifluoroacetic acid salt
Precaución: for Research Use Only


Formula: C₁₆H₂₀ClF₃N₄O₄
MDL Number: MFCD31536783
CAS Number: 1043444-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty