Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,4-Difluorophenyl)-3-(3-methyl-1-phenyl-1H-pyrazol-5-yl)urea 97%

Supplier: AMBEED, INC
Danger

A1209586-10MG A1209586-1MG A1209586-25MG A1209586-5MG
76677-938EA 53 USD InStock
76677-938 76677-940 76677-942 76677-944
1-(3,4-Difluorophenyl)-3-(3-methyl-1-phenyl-1H-pyrazol-5-yl)urea 97%
1-(3,4-Difluorophenyl)-3-(3-methyl-1-phenyl-1H-pyrazol-5-yl)urea
Precaución: for Research Use Only


Formula: C₁₇H₁₄F₂N₄O
MDL Number: MFCD27977571
CAS Number: 1443246-62-5
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty