Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

HQNO 97%

Supplier: AMBEED, INC
Warning

A1209614-10MG A1209614-1MG A1209614-50MG
76677-946EA 133 USD InStock
76677-946 76677-948 76677-950
HQNO 97%
HQNO
Precaución: for Research Use Only


Formula: C₁₆H₂₁NO₂
MDL Number: MFCD00057295
CAS Number: 341-88-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty