Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Chlorobenzyl)cyclopentanamine 97%

Supplier: AMBEED, INC
Danger

A1211262-250MG A1211262-5G
76677-994EA 14 USD InStock
76677-994 76677-996
N-(4-Chlorobenzyl)cyclopentanamine 97%
N-(4-Chlorobenzyl)cyclopentanamine
Precaución: for Research Use Only


Formula: C₁₂H₁₆ClN
MDL Number: MFCD03863988
CAS Number: 66063-15-8
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty