Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,2,2,3,3,4,4,4-Nonafluoro-N,N-bis(2-hydroxyethyl)butane-1-sulfonamide 98%

Supplier: AMBEED, INC
Danger

A1212417-25G A1212417-5G
76678-072EA 75 USD InStock
76678-072 76678-074
1,1,2,2,3,3,4,4,4-Nonafluoro-N,N-bis(2-hydroxyethyl)butane-1-sulfonamide 98%
1,1,2,2,3,3,4,4,4-Nonafluoro-N,N-bis(2-hydroxyethyl)butane-1-sulfonamide
Precaución: for Research Use Only


Formula: C₈H₁₀F₉NO₄S
MDL Number: MFCD32632476
CAS Number: 34455-00-0
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty