Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,4-Difluorophenyl)cyclopropanamine 98%

Supplier: AMBEED, INC
Warning

A1218096-100MG A1218096-1G A1218096-250MG
76678-334EA 49 USD InStock
76678-334 76678-336 76678-338
1-(3,4-Difluorophenyl)cyclopropanamine 98%
1-(3,4-Difluorophenyl)cyclopropanamine
Precaución: for Research Use Only


Formula: C₉H₉F₂N
MDL Number: MFCD09910057
CAS Number: 474709-85-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty