Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(1-Methyl-1H-pyrrol-2-ylmethylene)-4H-isoquinoline-1,3-dione 98%

Supplier: AMBEED, INC
Warning

A1219188-10MG A1219188-1MG A1219188-50MG A1219188-5MG
76678-366EA 47 USD InStock
76678-366 76678-368 76678-370 76678-372
4-(1-Methyl-1H-pyrrol-2-ylmethylene)-4H-isoquinoline-1,3-dione 98%
4-(1-Methyl-1H-pyrrol-2-ylmethylene)-4H-isoquinoline-1,3-dione
Precaución: for Research Use Only


Formula: C₁₅H₁₂N₂O₂
MDL Number: MFCD12912397
CAS Number: 1104546-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty