Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-(2,2-difluorobenzo[d][1,3]dioxol-5-yl)urea 98%

Supplier: AMBEED, INC
Warning

A1219208-10MG A1219208-25MG A1219208-5MG
76678-426EA 46 USD InStock
76678-426 76678-428 76678-430
1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-(2,2-difluorobenzo[d][1,3]dioxol-5-yl)urea 98%
1-(4-Chloro-3-(trifluoromethyl)phenyl)-3-(2,2-difluorobenzo[d][1,3]dioxol-5-yl)urea
Precaución: for Research Use Only


Formula: C₁₅H₈ClF₅N₂O₃
MDL Number: MFCD32667017
CAS Number: 2165324-62-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty