Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-(Thiophen-2-yl)phenyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A1219280-100MG A1219280-1G A1219280-250MG
76678-442EA 63 USD InStock
76678-442 76678-444 76678-446
2-(2-(Thiophen-2-yl)phenyl)acetic acid 97%
2-(2-(Thiophen-2-yl)phenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₀O₂S
MDL Number: MFCD20485723
CAS Number: 958219-45-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty