Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methoxyimino)propanoic acid 97%

Supplier: AMBEED, INC
Danger

A1219674-100MG A1219674-1G A1219674-250MG
76678-476EA 35 USD InStock
76678-476 76678-478 76678-480
2-(Methoxyimino)propanoic acid 97%
2-(Methoxyimino)propanoic acid
Precaución: for Research Use Only


Formula: C₄H₇NO₃
MW: 117.1033 g/mol
CAS Number: 27752-38-1
UN: 1325
ADR: 4.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty