Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-(2-(Bis(4-(trifluoromethyl)phenyl)phosphino)-5-(trifluoromethyl)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole 97%

Supplier: AMBEED, INC
Warning

A1219992-100MG A1219992-1G A1219992-250MG A1219992-50MG
76678-826EA 52 USD InStock
76678-826 76678-828 76678-830 76678-832
(R)-2-(2-(Bis(4-(trifluoromethyl)phenyl)phosphino)-5-(trifluoromethyl)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole 97%
(R)-2-(2-(Bis(4-(trifluoromethyl)phenyl)phosphino)-5-(trifluoromethyl)phenyl)-4-(tert-butyl)-4,5-dihydrooxazole
Precaución: for Research Use Only


Formula: C₂₈H₂₃F₉NOP
MW: 591.4477 g/mol
CAS Number: 1006708-91-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty