Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Bis(bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)ethane 97%

Supplier: AMBEED, INC
Warning

A1220061-100MG A1220061-1G A1220061-250MG
76678-948EA 86 USD InStock
76678-948 76678-950 76678-952
1,2-Bis(bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)ethane 97%
1,2-Bis(bis(3,5-bis(trifluoromethyl)phenyl)phosphanyl)ethane
Precaución: for Research Use Only


Formula: C₃₄H₁₆F₂₄P₂
MW: 942.4001 g/mol
MDL Number: MFCD01630832
CAS Number: 199342-62-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty