Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3aS,8aR)-2-(5-(Trifluoromethyl)pyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%

Supplier: AMBEED, INC
Warning

A1220224-100MG A1220224-1G A1220224-250MG
76679-136EA 39 USD InStock
76679-136 76679-138 76679-140
(3aS,8aR)-2-(5-(Trifluoromethyl)pyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole ≥97%, ee 99%
(3aS,8aR)-2-(5-(Trifluoromethyl)pyridin-2-yl)-3a,8a-dihydro-8H-indeno[1,2-d]oxazole
Precaución: for Research Use Only


Formula: C₁₆H₁₁F₃N₂O
CAS Number: 2412578-71-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty