Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,7-Dibromo-9-mesityl-10-methylacridin-10-ium tetrafluoroborate 97%

Supplier: AMBEED, INC
Warning

A1220243-100MG A1220243-1G A1220243-250MG A1220243-50MG
76679-154EA 88 USD InStock
76679-154 76679-156 76679-158 76679-160
2,7-Dibromo-9-mesityl-10-methylacridin-10-ium tetrafluoroborate 97%
2,7-Dibromo-9-mesityl-10-methylacridin-10-ium tetrafluoroborate
Precaución: for Research Use Only


Formula: C₂₃H₂₀BBr₂F₄N
MDL Number: MFCD32173816
CAS Number: 2635339-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty