Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(trans-2-hydroxycyclohexyl)-1H-indole-6-carboxylate 97%

Supplier: AMBEED, INC
Warning

A1220943-100MG A1220943-1G A1220943-250MG
76679-220EA 52 USD InStock
76679-220 76679-222 76679-224
Methyl 3-(trans-2-hydroxycyclohexyl)-1H-indole-6-carboxylate 97%
Methyl 3-(trans-2-hydroxycyclohexyl)-1H-indole-6-carboxylate
Precaución: for Research Use Only


Formula: C₁₆H₁₉NO₃
MDL Number: MFCD11111261
CAS Number: 936711-50-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty