Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,5'-(1,4-Phenylene)dipicolinaldehyde 98%

Supplier: AMBEED, INC
Warning

A1226399-100MG A1226399-1G A1226399-250MG
76679-402EA 126 USD InStock
76679-402 76679-404 76679-406
5,5'-(1,4-Phenylene)dipicolinaldehyde 98%
5,5'-(1,4-Phenylene)dipicolinaldehyde
Precaución: for Research Use Only


Formula: C₁₈H₁₂N₂O₂
MW: 288.3001 g/mol
MDL Number: MFCD32667053
CAS Number: 1399191-70-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty