Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4',4'',4'''-(Benzene-1,2,4,5-tetrayltetrakis(ethyne-2,1-diyl))tetraaniline 98%

Supplier: AMBEED, INC
Warning

A1226408-100MG A1226408-1G A1226408-250MG
76679-436EA 40 USD InStock
76679-436 76679-438 76679-440
4,4',4'',4'''-(Benzene-1,2,4,5-tetrayltetrakis(ethyne-2,1-diyl))tetraaniline 98%
4,4',4'',4'''-(Benzene-1,2,4,5-tetrayltetrakis(ethyne-2,1-diyl))tetraaniline
Precaución: for Research Use Only


Formula: C₃₈H₂₆N₄
MDL Number: MFCD32667057
CAS Number: 2375652-88-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty