Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(6-Amino-5-methylpyridin-3-yl)boronic acid 98%

Supplier: AMBEED, INC
Warning

A1227322-100MG A1227322-1G A1227322-250MG A1227322-50MG
76679-594EA 37 USD InStock
76679-594 76679-596 76679-598 76679-600
(6-Amino-5-methylpyridin-3-yl)boronic acid 98%
(6-Amino-5-methylpyridin-3-yl)boronic acid
Precaución: for Research Use Only


Formula: C₆H₉BN₂O₂
MW: 151.9589 g/mol
MDL Number: MFCD11865288
CAS Number: 1032759-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty