Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4-(methylamino)-4-oxobutanoic acid 98%

Supplier: AMBEED, INC
Warning

A1230378-1G A1230378-250MG A1230378-5G
76679-738EA 56 USD InStock
76679-738 76679-740 76679-742
2-Amino-4-(methylamino)-4-oxobutanoic acid 98%
2-Amino-4-(methylamino)-4-oxobutanoic acid
Precaución: for Research Use Only


Formula: C₅H₁₀N₂O₃
MW: 146.1445 g/mol
MDL Number: MFCD18914314
CAS Number: 757887-52-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty