Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluorophenyl trifluoromethanesulfonate 98%

Supplier: AMBEED, INC
Danger

A1239611-1G A1239611-25G A1239611-5G
76680-082EA 18 USD InStock
76680-082 76680-084 76680-086
3-Fluorophenyl trifluoromethanesulfonate 98%
3-Fluorophenyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₇H₄F₄O₃S
MDL Number: MFCD18974275
CAS Number: 57606-65-2
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty