Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-4-methylthiophene carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A1243530-100MG A1243530-1G A1243530-250MG
76680-128EA 57 USD InStock
76680-128 76680-130 76680-132
5-Chloro-4-methylthiophene carboxylic acid 98%
5-Chloro-4-methylthiophene carboxylic acid
Precaución: for Research Use Only


Formula: C₆H₅ClO₂S
MDL Number: MFCD01830083
CAS Number: 74598-03-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty