Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Benzyloxy-5-acetylisoxazole 98%

Supplier: AMBEED, INC
Warning

A1245886-100MG A1245886-1G A1245886-250MG
76680-154EA 50 USD InStock
76680-154 76680-156 76680-158
3-Benzyloxy-5-acetylisoxazole 98%
3-Benzyloxy-5-acetylisoxazole
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₃
MDL Number: MFCD25954749
CAS Number: 104164-47-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty