Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis(4-methyl-[1,1'-biphenyl]-3-yl)amine 98%

Supplier: AMBEED, INC
Warning

A1249918-1G A1249918-25G A1249918-5G
76680-258EA 10 USD InStock
76680-258 76680-260 76680-262
Bis(4-methyl-[1,1'-biphenyl]-3-yl)amine 98%
Bis(4-methyl-[1,1'-biphenyl]-3-yl)amine
Precaución: for Research Use Only


Formula: C₂₆H₂₃N
MDL Number: MFCD32671759
CAS Number: 1890112-76-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty