Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3aR,6R,7aR)-7,7-Dichloro-8,8-dimethylhexahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 99%

Supplier: AMBEED, INC
Warning

A1250068-1G A1250068-250MG A1250068-5G
76680-274EA 22 USD InStock
76680-274 76680-276 76680-278
(3aR,6R,7aR)-7,7-Dichloro-8,8-dimethylhexahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide 99%
(3aR,6R,7aR)-7,7-Dichloro-8,8-dimethylhexahydro-3H-3a,6-methanobenzo[c]isothiazole 2,2-dioxide
Precaución: for Research Use Only


Formula: C₁₀H₁₅Cl₂NO₂S
CAS Number: 204854-09-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty