Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(2-dicyclohexylphosphino-2',6'-diisopropoxy-1,1-biphenyl)[2-(2-aminoethylphenyl)]palladium(II) 2-methoxy-2-methylpropane complex 95%

Supplier: AMBEED, INC
Danger

A1251569-250MG A1251569-100MG A1251569-5G
76680-328EA 116 USD InStock
76680-328 77621-254 77621-256
Chloro(2-dicyclohexylphosphino-2',6'-diisopropoxy-1,1-biphenyl)[2-(2-aminoethylphenyl)]palladium(II) 2-methoxy-2-methylpropane complex 95%
Chloro(2-dicyclohexylphosphino-2',6'-diisopropoxy-1,1-biphenyl)[2-(2-aminoethylphenyl)]palladium(II) 2-methoxy-2-methylpropane complex
Precaución: for Research Use Only


Formula: C₄₃H₆₅ClNO₃PPd
MDL Number: MFCD12545955
CAS Number: 2361069-97-6
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty