Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Piperidin-4-ylboronic acid hydrochloride 98%

Supplier: AMBEED, INC
Warning

A1251601-1G A1251601-250MG A1251601-5G
76680-332EA 260 USD InStock
76680-332 76680-334 76680-336
Piperidin-4-ylboronic acid hydrochloride 98%
Piperidin-4-ylboronic acid hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₃BClNO₂
MW: 165.43 g/mol
MDL Number: MFCD11506080
CAS Number: 2306405-20-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty