Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(2,2′:6′,2″-terpyridine)platinum(II) chloride dihydrate 98%

Supplier: AMBEED, INC
Warning

A1256075-100MG A1256075-1G A1256075-250MG A1256075-50MG
76680-418EA 186 USD InStock
76680-418 76680-420 76680-422 76680-424
Chloro(2,2′:6′,2″-terpyridine)platinum(II) chloride dihydrate 98%
Chloro(2,2′:6′,2″-terpyridine)platinum(II) chloride dihydrate
Precaución: for Research Use Only


Formula: C₁₅H₁₅Cl₂N₃O₂Pt
MW: 535.2825 g/mol
MDL Number: MFCD00149950
CAS Number: 151120-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty