Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

10,11-Dimethoxy-6,8,13,13a-tetrahydro-5H-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinoline 98+%

Supplier: AMBEED, INC
Warning

A1258077-10MG A1258077-1MG A1258077-50MG A1258077-5MG
76680-532EA 33 USD InStock
76680-532 76680-534 76680-536 76680-538
10,11-Dimethoxy-6,8,13,13a-tetrahydro-5H-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinoline 98+%
10,11-Dimethoxy-6,8,13,13a-tetrahydro-5H-[1,3]dioxolo[4,5-g]isoquinolino[3,2-a]isoquinoline
Precaución: for Research Use Only


Formula: C₂₀H₂₁NO₄
CAS Number: 6656-19-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty